
Dodecamethylpentasiloxane

Linear Formula:
(CH3)3SiO[Si(CH3)2O]3Si(CH3)3
CAS Number:
Molecular Weight:
384.84
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Purity
Quality Segment
Reference Material Type
Special Grade
Showing 1-2 of 2