
(±)-Mevalonic acid 5-pyrophosphate tetralithium salt

Synonym(s):
(±)-MVAPP, (±)-Mevalonic acid 5-diphosphate tetralithium salt
Empirical Formula (Hill Notation):
C6H10Li4O10P2
CAS Number:
Molecular Weight:
331.85
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing