
Clomipramine impurity F

Synonym(s):
3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepine
Empirical Formula (Hill Notation):
C14H12ClN
CAS Number:
Molecular Weight:
229.70
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing