
Chiralyst P468

Synonym(s):
Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate
Empirical Formula (Hill Notation):
C17H24F3O3RhS
CAS Number:
Molecular Weight:
468.34
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing