
Apramycin sulfate salt

Synonym(s):
Nebramycin II
Empirical Formula (Hill Notation):
C21H41N5O11 · xH2SO4
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Activity Spectrum
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing