
AMOZ

Synonym(s):
3-Amino-5-morpholinomethyl-2-oxazolidinone
Empirical Formula (Hill Notation):
C8H15N3O3
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing