
Amodiaquine Hydrochloride Hydrate

Synonym(s):
Amodiaquin dihydrochloride dihydrate, 4-([7-Chloro-4-quinolinyl]amino)-2-([diethylamino]methyl)phenol
Empirical Formula (Hill Notation):
C20H22ClN3O · 2HCl · 2H2O
CAS Number:
Molecular Weight:
464.81
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing