
5-Bromoisophthalic acid

Synonym(s):
5-Bromo-1,3-benzenedicarbonic acid, 5-Bromo-1,3-benzenedicarboxylic acid, 5-Bromoisophthalic acid
Empirical Formula (Hill Notation):
C8H5BrO4
CAS Number:
Molecular Weight:
245.03
Products
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing