Skip to Content
Merck
4-Hydroxy-4-phenylpiperidine 99%

4-Hydroxy-4-phenylpiperidine

4-Hydroxy-4-phenylpiperidine 99%
Synonym(s):
4-Phenyl-4-piperidinol
Empirical Formula (Hill Notation):
C11H15NO
CAS Number:
Molecular Weight:
177.24
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing