
3-Methoxyphenylboronic acid

Synonym(s):
(m-Methoxyphenyl)boronic acid, 3-Methoxybenzeneboronic acid, m-Anisylboronic acid, m-Methoxybenzeneboronic acid
Linear Formula:
CH3OC6H4B(OH)2
CAS Number:
Molecular Weight:
151.96
Products
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing