2-Mercaptopyridine-3-carboxylic acid
Synonym(s):
2-Mercaptonicotinic acid
Empirical Formula (Hill Notation):
C6H5NO2S
CAS Number:
Molecular Weight:
155.17
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing