Skip to Content
Merck
All Photos(1)

Documents

PHL80473

(-)-Hydroxycitric acid lactone

phyproof® Reference Substance

Synonym(s):

(+)-Garcinia acid, (−)-Hydroxycitric acid lactone, (2S,3S)-3-Hydroxy-5-oxotetrahydrofuran-2,3-dicarboxylic acid, Garcinia lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O7
CAS Number:
Molecular Weight:
190.11
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

storage temp.

2-8°C

SMILES string

OC(=O)[C@H]1OC(=O)C[C@@]1(O)C(O)=O

InChI

1S/C6H6O7/c7-2-1-6(12,5(10)11)3(13-2)4(8)9/h3,12H,1H2,(H,8,9)(H,10,11)/t3-,6+/m1/s1

InChI key

PFHZIWAVXDSFTB-CVYQJGLWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by Phytolab GmbH & Co. KG

Application


  • Analysis of (-)-Hydroxycitric acid lactone in herbal products: A study employed capillary zone electrophoresis for the separation and analysis of (-)-Hydroxycitric acid and its lactone form, demonstrating its application in the quality control of herbal products containing Garcinia atroviridis Griff. This analytical method facilitates the accurate determination of active components in dietary supplements, supporting metabolic and enzyme inhibition studies (Muensritharam et al., 2008).

  • Quantitative analysis in Garcinia species: Research developed an improved liquid chromatographic method for the determination of organic acids including (-)-Hydroxycitric acid in various parts of Garcinia plants. This method is vital for metabolic pathway research, offering precise quantification crucial for studies related to weight loss and enzyme inhibition (Jayaprakasha et al., 2003).

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service