Skip to Content
Merck
All Photos(1)

Key Documents

SBR00021

Sigma-Aldrich

Nisin Ready Made Solution

(20,000-40,000 IU/mL in 0.02N HCl)

Synonym(s):

Nisaplin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C143H230N42O37S7
CAS Number:
Molecular Weight:
3354.07
UNSPSC Code:
12164500
NACRES:
NA.76

biological source

Lactococcus lactis

form

solution

concentration

(20,000-40,000 IU/mL in 0.02N HCl)

density

1.02 g/mL

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

cell membrane | interferes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(N[C@H](C(NC(C(N[C@@H](CCCCN)C(O)=O)=O)=C)=O)C(C)C)[C@@H](NC([C@H]([C@@H](C)CC)NC([C@@H](NC([C@@H](NC([C@H](CC1=CN=CN1)NC2=O)=O)CS[C@H]([C@@H]3C(N[C@H]2CS[C@H]([C@@H](NC([C@@H](NC([C@@H](NC([C@@H](NC([C@@H](NC(CNC([C@@H](NC([C@@H](NC([C@@H](NC(CNC4=O)=

InChI

1S/C143H230N42O37S7/c1-24-69(11)105(148)135(213)162-82(27-4)118(196)174-94-58-225-59-95(175-123(201)89(48-67(7)8)169-115(193)74(16)158-138(216)107(70(12)25-2)180-132(94)210)133(211)184-112-79(21)229-61-96(160-104(190)56-152-134(212)100-38-34-44-185(100)142(112)220)128(206)164-84(36-29-32-42-145)120(198)182-109-76(18)226-60-97(161-103(189)55-151-117(195)85(39-45-223-22)165-122(200)88(47-66(5)6)168-113(191)72(14)156-102(188)54-153-136(109)214)129(207)171-92(51-101(147)187)125(203)166-86(40-46-224-23)119(197)163-83(35-28-31-41-144)121(199)183-110-77(19)228-63-99-130(208)170-90(49-80-52-149-64-154-80)124(202)176-98(62-227-78(20)111(141(219)177-99)181-116(194)75(17)159-140(110)218)131(209)173-93(57-186)127(205)179-108(71(13)26-3)139(217)172-91(50-81-53-150-65-155-81)126(204)178-106(68(9)10)137(215)157-73(15)114(192)167-87(143(221)222)37-30-33-43-146/h27,52-53,64-72,75-79,83-100,105-112,186H,15-16,24-26,28-51,54-63,144-146,148H2,1-14,17-23H3,(H2,147,187)(H,149,154)(H,150,155)(H,151,195)(H,152,212)(H,153,214)(H,156,188)(H,157,215)(H,158,216)(H,159,218)(H,160,190)(H,161,189)(H,162,213)(H,163,197)(H,164,206)(H,165,200)(H,166,203)(H,167,192)(H,168,191)(H,169,193)(H,170,208)(H,171,207)(H,172,217)(H,173,209)(H,174,196)(H,175,201)(H,176,202)(H,177,219)(H,178,204)(H,179,205)(H,180,210)(H,181,194)(H,182,198)(H,183,199)(H,184,211)(H,221,222)/b82-27-

InChI key

NVNLLIYOARQCIX-GSJOZIGCSA-N

Application

Due to its wide range of antimicrobial activity, nisin is used as a shelf-life extender in a broad range of dairy and non-dairy products.

Biochem/physiol Actions

Nisin (Nisaplin) is produced by Lactococcus lactis. It is an effective lantibiotic peptide bactericidal agent. Nisin is a broad spectrum antimicrobial bacteriocin which affects Gram-positive bacteria, however, it has little or no effect on Gram-negative bacteria, yeasts or fungi.

It was demonstrated that nisin has capability to inhibit the growth of almost all tested Gram-positive beer-spoilage bacteria. Of 122 Gram-positive bacteria strains (predominantly Lactobacilli and Pediococci), 93% were sensitive to 100 units of nisin. In contrast, out of the 32 Gram-negative strains tested, only the genus Flavobacter proved to be affected by nisin.

Nisin exerts its antimicrobial activity by binding to lipid II which is a membrane-bound advanced intermediate involved in the biosynthesis of bacterial cell wall. By doing so, nisin sequesters lipid II from its functional location. In addition, the nisin-lipid II complex leads to formation of pores in the membrane causing cell death.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W-Y Cao et al.
Beneficial microbes, 11(1), 67-78 (2020-02-19)
Fibroblast growth factor 21 (FGF21), a metabolism regulator, has an important effect on metabolic diseases, such as obesity and diabetes. It is also expressed in mice, and the murine source has high homology with human FGF21. Recently, it has been
Lantibiotic nisin: Natural preservative from Lactococcus lactis
Suganthi, V.; et al.
International Research Journal of Pharmacy, 1, 13-19 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service