Skip to Content
Merck
All Photos(1)

Key Documents

KYSE-270

94072021, human esophagus, Epithelial

Synonym(s):

KY-270 Cells, KYSE270 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

KYSE-270, 94072021

biological source

human esophagus

growth mode

Adherent

karyotype

Hypotriploid with 16% polyploidy

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human oesophageal squamous cell carcinoma

Cell Line Description

Established from the well differentiated invasive oesophageal squamous cell carcinoma resected from the middle intra-thoracic oesophagus of a 79-year-old Japanese man prior to treatment. This cell line has been mycoplasma eradicated at ECACC.

Culture Medium

RPMI 1640:HAMS F12 (1:1) + 2mM Glutamine + 2% Foetal Bovine Serum (FBS)

Subculture Routine

Split confluent cultures 1:5 every second day using trypsin/EDTA;5% CO2; 37°C. Doubling time: 24 hours. Recommended seeding density: 2.5 x 10,000 cells/cm2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service