C8615
CXCL16 human
recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Synonym(s):
CXC chemokine ligand 16
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
Quality Level
recombinant
expressed in E. coli
Assay
≥97% (SDS-PAGE)
form
lyophilized powder
potency
2.5-12 ng/mL
mol wt
predicted mol wt ~10.1 kDa
packaging
pkg of 25 μg
storage condition
avoid repeated freeze/thaw cycles
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, tested
UniProt accession no.
storage temp.
−20°C
Physical form
Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 1.25 mg bovine serum albumin.
Analysis Note
The biological activity is measured by its ability to induce chemotaxis of mouse CXCR6 transfected BaF/3 cells.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Nature immunology, 1(4), 298-304 (2001-03-23)
We describe a protein with the hallmarks of a chemokine, designated CXCL16, that is made by dendritic cells (DCs) in lymphoid organ T cell zones and by cells in the splenic red pulp. CXCL16 contains a transmembrane domain and both
Oncotarget, 6(12), 9985-9998 (2015-04-19)
Lung cancer (LuCa) is the leading cause of cancer-related deaths worldwide regardless of the gender. High mortality associated with LuCa is due to metastasis, molecular mechanisms of which are yet to be defined. Here, we present evidence that chemokine receptor
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service