Skip to Content
Merck
All Photos(1)

Documents

Y0000492

Oxycodone hydrochloride

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21NO4 · HCl
CAS Number:
Molecular Weight:
351.82
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

oxycodone

manufacturer/tradename

EDQM

drug control

USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.[H][C@@]12Oc3c(OC)ccc4C[C@H]5N(C)CC[C@@]1(c34)[C@@]5(O)CCC2=O

InChI

1S/C18H21NO4.ClH/c1-19-8-7-17-14-10-3-4-12(22-2)15(14)23-16(17)11(20)5-6-18(17,21)13(19)9-10;/h3-4,13,16,21H,5-9H2,1-2H3;1H/t13-,16+,17+,18-;/m1./s1

InChI key

MUZQPDBAOYKNLO-RKXJKUSZSA-N

Gene Information

human ... OPRM1(4988)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Oxycodone hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

μ and κ opioid receptor agonist.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caitlin M Vander Weele et al.
The European journal of neuroscience, 40(7), 3041-3054 (2014-09-12)
While most drugs of abuse increase dopamine neurotransmission, rapid neurochemical measurements show that different drugs evoke distinct dopamine release patterns within the nucleus accumbens. Rapid changes in dopamine concentration following psychostimulant administration have been well studied; however, such changes have
Salvatore V Colucci et al.
Clinical drug investigation, 34(6), 421-429 (2014-04-24)
Abuse of opioid analgesics has become a public health issue. Some opioid abusers use intravenous administration to increase the magnitude of positive reinforcing effects. Intravenous co-administration of oxycodone with naloxone, an opioid antagonist, may reduce these rewarding effects and discourage
Carrie L Wade et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 40(2), 421-428 (2014-07-26)
The abuse of prescription opioids that are used for the treatment of chronic pain is a major public health concern, costing ∼$53.4 billion annually in lost wages, health-care costs, and criminal costs. Although opioids remain a first-line therapy for the
Jason A Miranda et al.
PloS one, 9(8), e106108-e106108 (2014-08-27)
Sensory processing in the spinal cord during disease states can reveal mechanisms for novel treatments, yet very little is known about pain processing at this level in the most commonly used animal models of articular pain. Here we report a
Tomoe Kanbara et al.
Neuroscience letters, 580, 119-124 (2014-08-17)
It has begun to be understood that μ-opioid receptor (MOR) produces ligand-biased agonism, which contributes to differential physiological functions of MOR agonists. We previously demonstrated that in oxaliplatin-induced neuropathy in rats, morphine and oxycodone exhibited antinociceptive effects while antinociception of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service