Skip to Content
Merck
All Photos(2)

Key Documents

909556

Sigma-Aldrich

Rhodium(III) acetate

Umicore

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Rh(OAc)3
CAS Number:
UNSPSC Code:
11101711
NACRES:
NA.22

form

powder

mp

238 °C (decomp)

InChI

1S/C2H4O2.Rh/c1-2(3)4;/h1H3,(H,3,4);/p-1

InChI key

NENDHUHGFRLXEN-UHFFFAOYSA-M

Application

Rhodium(III) acetate is a homogeneous catalyst useful for oxidation reactions.

Packaging

Do not store above 25°C (77°F). Store in a tightly closed original container under dry inert gas, protected from direct sunlight in a dry, cool and well-ventilated area.

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service