Skip to Content
Merck
All Photos(4)

Documents

MAB1618

Sigma-Aldrich

Anti-Dynein Antibody, 74 kDa Intermediate chains, cytoplasmic, clone 74.1

clone 74.1, Chemicon®, from mouse

Synonym(s):

Cytoplasmic dynein 1 intermediate chain 1, Cytoplasmic dynein intermediate chain 1, Dynein intermediate chain 1, cytosolic, DH IC-1, Cytoplasmic dynein 1 intermediate chain 2, Cytoplasmic dynein intermediate chain 2, Dynein intermediate chain 2, cytosoli

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

74.1, monoclonal

species reactivity

Xenopus, mouse, fish, rat, human, sheep, Drosophila, bovine

should not react with

squid

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DYNC1I1(1780)

General description

Cytoplasmic dynein 1 intermediate chain 1 (UniProt Q29RQ3; also known as Cytoplasmic dynein intermediate chain 1, Dynein intermediate chain 1, cytosolic, DH IC-1) and Cytoplasmic dynein 1 intermediate chain 2 (UniProt Q0III3; also known as Cytoplasmic dynein intermediate chain 2, Dynein intermediate chain 2, cytosolic, DH IC-2) are encoded by the DYNC1I1 gene (Gene ID 613724) and DYNC1I2 gene (Gene ID 526329) in bovine, respectively. Cytoplasmic dynein complexes are microtubules-associated retrograde transport motors. Cytoplasmic dynein 1 is the more abundant motor in cells, while cytoplasmic dynein 2 takes part in intraflagellar transport. Cytoplasmic dynein 1 is a multisubunit complex of ∼1.5 MDa composed of a homodimer of heavy chains (encoded by DYNC1H1), two intermediate chains (encoded by DYNC1I1 and DYNC1I2), light-intermediate chains (encoded by DYNC1LI1, DYNC1LI2) and light chains (encoded by DYNLT1, DYNLT3, DYNLRB1, DYNLRB2, DYNLL1, DYNLLl2). The heavy chain homodimer acts as the dynein complex core and binds microtubules to enable ATP-dependent cytoplasmic dynein movement. The other dynein subunits associate as homodimers and play regulatory roles in maintaining the complex stability, modulating its activity, and mediating its interaction with accessory and cargo proteins. The intermediate chain proteins (IC1 & IC2) are involved in cargo binding and specificity, either alone or via interaction with the dynactin complex. IC1 and IC2 share 69% protein identity. They interact with the dynein light chains and the p150 subunit of dynactin at the N-terminus and with the heavy chains through WD40 repeats at the intermediate chain C-terminus.

Specificity

Not tested on chicken, nematodes or Dictystelium.

Application

Anti-Dynein Antibody, 74 kDa Intermediate chains, cytoplasmic, clone 74.1 detects level of Dynein & has been published & validated for use in IC, IF, IP & WB.
Immunocytochemistry:
1:50-1:100 dilution from a previous lot was used. Reacts with cultured MDCK cells, NRR cells, N1E cells, and PTK-1 cells.

Immunoprecipitation:
10 μL of a previous lot per 0.5 gm tissue was used. The monoclonal cleanly immunoprecipitates the entire dynein complex stoichiometrically from TX-100 or NP-40 lysates (including the 530 kD heavy chain, the light intermediate chains and the light chains) from various tissues and cultured cell lines. If detergents such as SDS are used to prepare the lysates the only dynein subunits which are immunoprecipitated are the IC74 subunits (presumably because the dynein complex dissociates), and other contaminating proteins can be found in SDS immunoprecipitates.

Immunoblotting:
1:1,000-1:5000. Reacts with cultured MDCK cells, PC-12 cells, N1E cells, neurons, glia and other cultured cells.

Optimal working dilutions must be determined by end user.

Quality

Routinely evaluated by Western Blot on A431 lysates.

Western Blot Analysis:
1:500 dilution of this lot detected DYNEIN on 10 μg of A431 lysates.

Target description

~74 kDa observed.

Physical form

Format: Purified
Purified mouse monoclonal IgG2b in buffer containing 20 mM sodium phosphate, 250 mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative.

Analysis Note

Control
HeLa cells, A431 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dileep Varma et al.
The Journal of cell biology, 182(6), 1045-1054 (2008-09-24)
Cytoplasmic dynein has been implicated in diverse mitotic functions, several involving its association with kinetochores. Much of the supporting evidence comes from inhibition of dynein regulatory factors. To obtain direct insight into kinetochore dynein function, we expressed a series of
Hsp90-binding immunophilins link p53 to dynein during p53 transport to the nucleus.
Galigniana, MD; Harrell, JM; O'Hagen, HM; Ljungman, M; Pratt, WB
The Journal of Biological Chemistry null
Jen-Hsuan Wei et al.
The Journal of cell biology, 184(3), 391-397 (2009-02-04)
The mammalian Golgi ribbon disassembles during mitosis and reforms in both daughter cells after division. Mitotic Golgi membranes concentrate around the spindle poles, suggesting that the spindle may control Golgi partitioning. To test this, cells were induced to divide asymmetrically
Daniël Splinter et al.
Molecular biology of the cell, 23(21), 4226-4241 (2012-09-08)
Cytoplasmic dynein is the major microtubule minus-end-directed cellular motor. Most dynein activities require dynactin, but the mechanisms regulating cargo-dependent dynein-dynactin interaction are poorly understood. In this study, we focus on dynein-dynactin recruitment to cargo by the conserved motor adaptor Bicaudal
Shiqiao Ye et al.
PloS one, 6(11), e27285-e27285 (2011-11-11)
Microtubule organization and lysosomal secretion are both critical for the activation and function of osteoclasts, highly specialized polykaryons that are responsible for bone resorption and skeletal homeostasis. Here, we have identified a novel interaction between microtubule regulator LIS1 and Plekhm1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service