Skip to Content
Merck
All Photos(2)

Key Documents

ABS1486

Sigma-Aldrich

Anti-E3 ubiquitin-protein ligase RNF31

from rabbit, purified by affinity chromatography

Synonym(s):

EC: 2.3.2.27, HOIL-1-interacting protein, HOIP, RING finger protein 31, RING-type E3 ubiquitin transferase RNF31, Zinc in-between-RING-finger ubiquitin-associated domain protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

chimpanzee ... Rnf31(452810)
human ... RNF31(55072)

General description

E3 ubiquitin-protein ligase RNF31 (UniProt: Q96EP0; also known as EC: 2.3.2.27, HOIL-1-interacting protein, HOIP, RING finger protein 31, RING-type E3 ubiquitin transferase RNF31, Zinc in-between-RING-finger ubiquitin-associated domain protein) is encoded by the RNF31 (also known as ZIBRA) gene (Gene ID: 55072) in human. E3 ubiquitin-protein ligase RNF31 is a cytoplasmic protein that is a component of the linear ubiquitin chain assembly complex (LUBAC complex), which conjugates linear (′Met-1′-linked) polyubiquitin chains to substrates and plays a key role in NF-kappa-B activation and regulation of inflammation. LUBAC conjugates linear polyubiquitin to IKBKG (NEMO) and receptor interacting serine/threonine Kinase 1 (RIPK1) and is involved in activation of the canonical NF-kappa-B and the JNK signaling pathways. LUBAC-mediated polyubiquitination interferes with TNF-induced cell death and thereby prevents inflammation. Together with OUTLIN deubiquitinase, the LUBAC complex is also reported to regulate the canonical Wnt signaling pathway during angiogenesis. E3 ubiquitin-protein ligase RNF31 is expressed in both normal and transformed breast epithelial cell lines. It contains a polyubiquitin-binding (PUB) domain (aa 71-142) that mediates interaction with the PCNA-interacting motifs of C-terminus of the AAA+ ATPase p97 (VCP) and RNF31, with a strong preference for RNF3. It also has a ubiquitin-associated (UBA) domain (aa 564-615) that mediates association with RanBP-type and C3HC4-type zinc finger-containing protein 1 (RBCK1/HOIL1) via interaction with its ubiquitin-like (UBL) domain. (Ref.: Elliot, PR et al (2014). Mol. Cell 54(3); 335-48).

Specificity

This rabbit polyclonal antibody detects E3 ubiquitin-protein ligase RNF31 in human cells. It targets an epitope within 17 amino acids from the C-terminal region. Immunogen sequence is conserved in all 3 isoforms.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to 17 amino acids from the C-terminal region of human E3 ubiquitin-protein ligase RNF31.

Application

Anti-E3 ubiquitin-protein ligase RNF31, Cat. No. ABS1486, is a rabbit polyclonal antibody that detects E3 ubiquitin-protein ligase RNF31 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected E3 ubiquitin-protein ligase RNF31 in human adrenal gland and human uterus tissues.
Research Category
Signaling

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected E3 ubiquitin-protein ligase RNF31 in 10 µg of Jurkat cell lysate.

Target description

~120 kDa observed; 119.65 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service