Skip to Content
Merck
All Photos(1)

Key Documents

5.32336

Sigma-Aldrich

NOX Inhibitor VIII, VAS3947

Synonym(s):

NOX Inhibitor VIII, VAS3947, 3-Benzyl-7-(1,3-oxazol-2-ylsulfanyl)-3H-[1,2,3]triazolo[4,5-d]pyrimidine, 2-((3-Benzyl-3H-[1,2,3]triazolo[4,5-d]pyrimidin-7-yl)thio)oxazole, NADPH Oxidase Inhibitor VIII, VAS-3947

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10N6OS
CAS Number:
Molecular Weight:
310.33
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable VAS2870 (Cat No. 492000) analogue that displays improved solubility and effectively blocks ROS production in DMSO-differentiated HL-60 cultures upon oxidative burst induction by PMA (IC50 = 1 µM by Cytochrome C reduction & 2 µM by L012 chemiluminescence; Drug added 30 min prior to 100 nM PMA/500582 & 524400) via NADPH oxidases/NOX inhibition, while displaying no potency against eNOS or xanthine oxidase/XOD enzymatic activity in cell-free assays even at a high concentration of 100 µM. Reported to suppress the high ROS level in isolated thoracis aortas sections from spontaneously hypertensive rats (SHRs) in situ (by 66% with 10 µM) and synergize with Imatinib (Cat. No. 504595) against the proliferation of p210 Bcr-abl-expressing murine Boff 210 cultures. A commonly employed NOX inhibitor Apocynin (Cat. No. 178385), in comparison, is shown to interfere with L012-based assays and exhibit little potency against NOX-mediated ROS production in DMSO-differentiated HL-60 cultures (by Cytochrome C reduction) or in cell-free assays using A7r5 & CaCo-2 homogenates (IC50 >1 mM), while another NOX inhibitor DPI (Cat. No. 300260) is shown to inhibit both eNOS & XOD activities.
A cell-permeable triazolo-pyrimidine derivative of VAS2870 (Cat. No. 492000) that displays enhanced solubility and acts as a selective inhibitor NOX family of NADPH oxidase. Shown to inhibit NOX activity in CaCo-2 cells (expressing NOX 1, 2, 4, and 5; IC50 = 12 µM), HL-60 cells (mainly expressing NOX2; IC50 = 2 µM), and A7r5 cells (mainly expressing NOX3 and 4; IC50 = 13 µM). Does not affect the activities of xanthine oxidase and endothelial nitric oxide synthase (eNOS) even at higher concentrations (~100 µM). Acts synergistically with Imatinib (Cat. No. 504595) to reduce the proliferation of chronic myeloid leukemia (CML) cells.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
NADPH Oxidase

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Sanchez-Sanchez, B., et al. 2014. Clin. Cancer Res.20, 4014.
Altenhofer, S., et al., 2012. Cell. Mol. Life Sci.69, 2327.
Wind, S., et al. 2010. Br. J. Pharmacol.161, 885.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service