Skip to Content
Merck
All Photos(1)

Key Documents

361565

Sigma-Aldrich

GSK3β Inhibitor XIX, IM-12

The GSK3β Inhibitor XIX, IM-12 controls the biological activity of GSK3β. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

GSK3β Inhibitor XIX, IM-12, IM-12

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20FN3O2
Molecular Weight:
377.41
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange-yellow

solubility

DMSO: 100 mg/mL
DMSO: soluble

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable indolylmaleimide derivative and SB-216763 analog that acts as a GSK-3β inhibitor, with an IC50 of 53 nM in an in vitro binding assay, compared with 92 nM for SB-216763 in the same assay condition, and reduces GSK-3β activity to 27% in an enzymatic assay at 3 µM concentration. It is shown to attenuate hNPC (human neural progenitor cells) cell proliferation, and enrich β-catenin around the nucleus of ST14 cells in the same extent of SB-216763. Furthermore, it can mimic the effect of stabilized β-catenin, up-regulates TCF-activity in a luciferase reporter gene assay, and enhances human progenitor cell differentiation.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Schmole, AC., et al. 2010. Bioorg. Med. Chem.18, 6785.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service