Skip to Content
Merck
All Photos(1)

Key Documents

1.93623

Supelco

Zinc chloride

EMPLURA®

Synonym(s):

Zinc chloride, Dichlorozinc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnCl2
CAS Number:
Molecular Weight:
136.30
MDL number:
UNSPSC Code:
12352302

vapor pressure

1 mmHg ( 428 °C)

product line

EMPLURA®

mp

293 °C (lit.)

storage temp.

20-25°C

SMILES string

[Cl-].[Cl-].[Zn++]

InChI

1S/2ClH.Zn/h2*1H;/q;;+2/p-2

InChI key

JIAARYAFYJHUJI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service