Skip to Content
Merck
All Photos(1)

Key Documents

12-121

Sigma-Aldrich

PKC Inhibitor Peptide

Sign Into View Organizational & Contract Pricing


About This Item

eCl@ss:
32160405

description

protein target PKC

technique(s)

activity assay: suitable

UniProt accession no.

General description

FUNCTION: SwissProt: P17252 # PKC is activated by diacylglycerol which in turn phosphorylates a range of cellular proteins. PKC also serves as the receptor for phorbol esters, a class of tumor promoters.
COFACTOR: Binds 3 calcium ions per subunit. The ions are bound to the C2 domain (By similarity).
SIZE: 672 amino acids; 76764 Da
SUBUNIT: Interacts with CENTA1, CSPG4 and PRKCABP. Binds to SDPR in the presence of phosphatidylserine.
SIMILARITY: SwissProt: P17252 ## Belongs to the protein kinase superfamily. AGC Ser/Thr protein kinase family. PKC subfamily. & Contains 1 AGC-kinase C-terminal domain. & Contains 1 C2 domain. & Contains 2 phorbol-ester/DAG-type zinc fingers. & Contains 1 protein kinase domain.

Quality

Routinely evaluated by inhibition of Protein Kinase C

Physical form

Lyophilized powder

Storage and Stability

1 year at -20°C

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service