Skip to Content
Merck
All Photos(1)

Key Documents

JKA7068

Sigma-Aldrich

4arm-PEG10K

average Mn 10,000, Acrylate

Synonym(s):

4arm-PEG-Acrylate, 4arm-PEG-ACLT, Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C(CH2O(CH2CH2O)nCH2CH2OOCCHCH2)4
UNSPSC Code:
12162002
NACRES:
NA.23

product name

4arm-PEG10K-Acrylate, average Mn 10,000

form

solid

mol wt

average Mn 10,000

reaction suitability

reaction type: Polymerization Reactions

polymer architecture

shape: 4-arm
functionality: homofunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

4arm PEG Acrylate. Hydrogel PEG. Used in vinyl polymerization or co-polymerization.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Schukur, L., et al.
Advanced Helathcare Materials, 2(1), 195-205 (2013)

Articles

Highlighting new synthetic modifications of PEG to improve the mechanical properties and degradation of resulting hydrogels in tissue engineering applications.

Highlighting existing and novel fabrication methods for both, solid and hydrogel-based scaffold for tissue engineering applications.

Professor Shrike Zhang (Harvard Medical School, USA) discusses advances in 3D-bioprinted tissue models for in vitro drug testing, reviews bioink selections, and provides application examples of 3D bioprinting in tissue model biofabrication.

Professor Shrike Zhang (Harvard Medical School, USA) discusses advances in 3D-bioprinted tissue models for in vitro drug testing, reviews bioink selections, and provides application examples of 3D bioprinting in tissue model biofabrication.

See All

Protocols

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service