Skip to Content
Merck
All Photos(1)

Key Documents

767352

Sigma-Aldrich

Stearic acid N-hydroxysuccinimide ester

Synonym(s):

NHS protected stearate, NHS-octadecanoic acid, Octadecanoic acid N-hydroxysuccinimide ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H39NO4
CAS Number:
Molecular Weight:
381.55
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

solid

mp

71-76 °C

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)ON1C(=O)CCC1=O

InChI

1S/C22H39NO4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-22(26)27-23-20(24)18-19-21(23)25/h2-19H2,1H3

InChI key

ZERWDZDNDJBYKA-UHFFFAOYSA-N

General description

Stearic acid N-hydroxysuccinimide ester is an N-Hydroxysuccinimide ester of fatty acid. They are used in the synthesis of N-acyl amino acids and fatty acyl CoA. They are also used in the direct N-acylation of sphingenine or sphinganine thus leading to the formation of ceramides.

Application

Used in the synthesis of ceramides and in the hydrophobication of amines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D E Ong et al.
Journal of lipid research, 13(6), 819-822 (1972-11-01)
Fatty acyl esters of N-hydroxysuccinimide have been used to directly N-acylate sphingenine or sphinganine, forming the corresponding ceramides. The reaction proceeds in excellent yield (84-96%) from small amounts of starting material (10-20 mg). The product ceramides are pure after one

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service