Skip to Content
Merck
All Photos(1)

Key Documents

745529

Sigma-Aldrich

4-Heptyl-4-13C acetate

99 atom % 13C, 98% (CP)

Synonym(s):

13C Labeled 4-Heptyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC8H18O2
Molecular Weight:
159.23
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

liquid

mass shift

M+1

SMILES string

CCC[13CH](OC(C)=O)CCC

InChI

1S/C9H18O2/c1-4-6-9(7-5-2)11-8(3)10/h9H,4-7H2,1-3H3/i9+1

InChI key

JPXGPRBLTIYFQG-QBZHADDCSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.4 °F

Flash Point(C)

68 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service