HPLC Analysis of HSA and HSA Fusion Protein on Zenix®-C SEC-300
Materials
analytical column
Product No.
Description
Pricing
Zenix®-C SEC-300 Gel Filtration Column, 3 μm
L × I.D. 30 cm × 7.8 mm, 3 μm particle size, stainless steelCONDITIONS
column
Zenix-C SEC-300, 30 cm x 7.8 mm I.D., 3 μm particles (Z777092)
mobile phase
(A) 150 mM sodium phosphate buffer, pH 7.0; (B) IPA; (95:5, v/v, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 280 nm
injection
10 μL
sample
HSA fusion peptide 5 mg/mL (MW 75 kD, pl 5.0, HSA conjugated peptide in diabetes treatment)
Description
Legal Information
Zenix is a registered trademark of Sepax Technologies