Merck
  • The IKK/NF-κB signaling pathway requires Morgana to drive breast cancer metastasis.
The IKK/NF-κB signaling pathway requires Morgana to drive breast cancer metastasis.
Nature communications (2017-11-22)
Federica Fusella, Laura Seclì, Elena Busso, Anna Krepelova, Enrico Moiso, Stefania Rocca, Laura Conti, Laura Annaratone, Cristina Rubinetto, Maurizia Mello-Grand, Vijay Singh, Giovanna Chiorino, Lorenzo Silengo, Fiorella Altruda, Emilia Turco, Alessandro Morotti, Salvatore Oliviero, Isabella Castellano, Federica Cavallo, Paolo Provero, Guido Tarone, Mara Brancaccio
ABSTRACT
NF-κB is a transcription factor involved in the regulation of multiple physiological and pathological cellular processes, including inflammation, cell survival, proliferation, and cancer cell metastasis. NF-κB is frequently hyperactivated in several cancers, including triple-negative breast cancer. Here we show that NF-κB activation in breast cancer cells depends on the presence of the CHORDC1 gene product Morgana, a previously unknown component of the IKK complex and essential for IκBα substrate recognition. Morgana silencing blocks metastasis formation in breast cancer mouse models and this phenotype is reverted by IκBα downregulation. High Morgana expression levels in cancer cells decrease recruitment of natural killer cells in the first phases of tumor growth and induce the expression of cytokines able to attract neutrophils in the primary tumor, as well as in the pre-metastatic lungs, fueling cancer metastasis. In accordance, high Morgana levels positively correlate with NF-κB target gene expression and poor prognosis in human patients.
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
Anti-Vinculin antibody, Mouse monoclonal, clone V284, purified from hybridoma cell culture
Sigma-Aldrich
Anti-CHORDC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
MISSION® esiRNA, targeting human CDC37
Sigma-Aldrich
Maltose solution, for molecular biology, BioReagent, ~20% in H2O
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
MISSION® esiRNA, targeting human CHORDC1