Merck
  • Characterization of a new adenovirus isolated from black-tailed deer in California.
Characterization of a new adenovirus isolated from black-tailed deer in California.
Archives of virology (2001-08-16)
H D Lehmkuhl, L A Hobbs, L W Woods
ABSTRACT
An adenovirus associated with systemic and localized vascular damage was demonstrated by transmission electron microscopy and immunohistochemistry in a newly recognized epizootic hemorrhagic disease in California black-tailed deer. In this study, we describe the cultural, physicochemical and serological characteristics of a virus isolated from lung using neonatal white-tail deer lung and turbinate cell cultures. The virus had the cultural, morphological and physicochemical characteristics of members of the Adenoviridae family. The virus would not replicate in low passage fetal bovine, caprine or ovine cells. Antiserum to the deer adenovirus, strain D94-2569, neutralized bovine adenovirus type-6 (BAdV-6), BAdV-7, and caprine adenovirus type-1 (GAdV-1). Antiserum to BAdV-6 did not neutralize the deer adenovirus but antiserum to BAdV-7 and GAdV-1 neutralized the deer adenovirus. Cross-neutralization with the other bovine, caprine and ovine adenovirus species was not observed. Restriction endonuclease patterns generated for the deer adenovirus were unique compared to those for the currently recognized bovine, caprine and ovine adenovirus types. Amino acid sequence alignments of the hexon gene from the deer adenovirus strain D94-2569 indicate that it is a member of the proposed new genus (Atadenovirus) of the Adenoviridae family. While closely related antigenically to BAdV-7 and GAdV-1, the deer adenovirus appears sufficiently distinct culturally and molecularly to justify consideration as a new adenovirus type.
Product Description
G-CAPSULE electroelution device
G-CAPSULE Accessory Weight