Merck
  • Degradation of transcription factor RFX5 during the inhibition of both constitutive and interferon gamma-inducible major histocompatibility complex class I expression in chlamydia-infected cells.
Degradation of transcription factor RFX5 during the inhibition of both constitutive and interferon gamma-inducible major histocompatibility complex class I expression in chlamydia-infected cells.
The Journal of experimental medicine (2000-05-03)
G Zhong, L Liu, T Fan, P Fan, H Ji
ABSTRACT
We have previously shown that the obligate intracellular pathogen chlamydia can suppress interferon (IFN)-gamma-inducible major histocompatibility complex (MHC) class II expression in infected cells by degrading upstream stimulation factor (USF)-1. We now report that chlamydia can also inhibit both constitutive and IFN-gamma-inducible MHC class I expression in the infected cells. The inhibition of MHC class I molecule expression correlates well with degradation of RFX5, an essential downstream transcription factor required for both the constitutive and IFN-gamma-inducible MHC class I expression. We further demonstrate that a lactacystin-sensitive proteasome-like activity identified in chlamydia-infected cell cytosolic fraction can degrade both USF-1 and RFX5. This proteasome-like activity is dependent on chlamydial but not host protein synthesis. Host preexisting proteasomes may not be required for the unique proteasome-like activity. These observations suggest that chlamydia-secreted factors may directly participate in the proteasome-like activity. Efforts to identify the chlamydial factors are underway. These findings provide novel information on the molecular mechanisms of chlamydial evasion of host immune recognition.
Sigma-Aldrich
MG-132, A cell-permeable, potent, reversible proteasome inhibitor (Ki = 4 nM).
Sigma-Aldrich
clasto-Lactacystin β-Lactone, A highly specific, cell-permeable, and irreversible 20S proteasome inhibitor.
Sigma-Aldrich
Anti-20S proteasome subunit α2 Antibody, clone MCP21, clone MCP21, Upstate®, from mouse
Sigma-Aldrich
MG-115, Potent reversible proteasome inhibitor (Ki = 21 nM and 35 nM for 20S and 26S proteasome, respectively).