Merck
  • Osteopontin Exacerbates Pulmonary Damage in Influenza-Induced Lung Injury.
Osteopontin Exacerbates Pulmonary Damage in Influenza-Induced Lung Injury.
Japanese journal of infectious diseases (2015-04-14)
Yunan Zhu, Yingfeng Wei, Jianing Chen, Guangying Cui, Yulong Ding, Masashi Kohanawa, Xuejun Xu, Hongyan Diao
ABSTRACT
The level of osteopontin (OPN) increases during bacterial lung infection. However, the OPN level in virus-induced lung injury is unclear, and the relationship between the hyer-production of OPN and lung injury remains to be thoroughly understood. Therefore, we sought to determine whether a relationship exists between OPN and pulmonary damage. Particularly, pulmonary edema and the destruction of pulmonary tissue. In this study, we found that the OPN level was significantly elevated in patients with pulmonary damage, and there was a positive correlation between the OPN serum level and disease severity in influenza lung injury. The epithelial sodium channel (ENaC) is the main mechanism of clearance of pulmonary edema fluid, and matrix metalloproteinase 7 (MMP7) can degrade the extracellular matrix. In lung epithelial cells, OPN markedly decreased the mRNA expression of the α-subunit of ENaC through integrin β3 and CD44 (OPN receptors); however, the expression of MMP7 was promoted by OPN interaction with integrin β1 and CD44. In addition, OPN increased the levels of tumor necrosis factor-α and interleukin-6. These findings suggested that OPN might increase influenza virus-induced lung injury by augmenting lung epithelial cell apoptosis and impairing ENaC and extracellular matrix destruction.
Sigma-Aldrich
Osteopontin from mouse, >95% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Osteopontin human, recombinant, expressed in NSO cells, ≥95% (SDS-PAGE), lyophilized powder, suitable for cell culture
Sigma-Aldrich
Osteopontin human, recombinant, expressed in HEK 293 cells, ≥97% (SDS-PAGE), ≥97% (HPLC), suitable for cell culture