Merck
  • Towards a better understanding on agglomeration mechanisms and thermodynamic properties of TiO₂ nanoparticles interacting with natural organic matter.
Towards a better understanding on agglomeration mechanisms and thermodynamic properties of TiO₂ nanoparticles interacting with natural organic matter.
Water research (2015-05-23)
Frédéric Loosli, Letícia Vitorazi, Jean-François Berret, Serge Stoll
ABSTRACT
Interaction between engineered nanoparticles and natural organic matter is investigated by measuring the exchanged heat during binding process with isothermal titration calorimetry. TiO2 anatase nanoparticles and alginate are used as engineered nanoparticles and natural organic matter to get an insight into the thermodynamic association properties and mechanisms of adsorption and agglomeration. Changes of enthalpy, entropy and total free energy, reaction stoichiometry and affinity binding constant are determined or calculated at a pH value where the TiO2 nanoparticles surface charge is positive and the alginate exhibits a negative structural charge. Our results indicate that strong TiO2-alginate interactions are essentially entropy driven and enthalpically favorable with exothermic binding reactions. The reaction stoichiometry and entropy gain are also found dependent on the mixing order. Finally correlation is established between the binding enthalpy, the reaction stoichiometry and the zeta potential values determined by electrophoretic mobility measurements. From these results two types of agglomeration mechanisms are proposed depending on the mixing order. Addition of alginate in TiO2 dispersions is found to form agglomerates due to polymer bridging whereas addition of TiO2 in alginate promotes a more individually coating of the nanoparticles.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, 1 M
Sigma-Aldrich
Sodium hydroxide solution, 0.01 M
Sigma-Aldrich
Sodium hydroxide solution, 0.1 M
Sigma-Aldrich
Sodium hydroxide solution, 7 M
Sigma-Aldrich
Sodium hydroxide solution, 4 M
Sigma-Aldrich
Sodium hydroxide solution, 0.05 M
Sigma-Aldrich
Sodium hydroxide, JIS special grade, ≥96.0%
Sigma-Aldrich
Sodium hydroxide solution, 6 M
Sigma-Aldrich
Sodium hydroxide solution, 0.2 M
Sigma-Aldrich
Sodium hydroxide, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis