Merck
  • Lipoxin A4 Activates Nrf2 Pathway and Ameliorates Cell Damage in Cultured Cortical Astrocytes Exposed to Oxygen-Glucose Deprivation/Reperfusion Insults.
Lipoxin A4 Activates Nrf2 Pathway and Ameliorates Cell Damage in Cultured Cortical Astrocytes Exposed to Oxygen-Glucose Deprivation/Reperfusion Insults.
Journal of molecular neuroscience : MN (2015-02-24)
Le Wu, Hong-Hua Li, Qiang Wu, Sen Miao, Zi-Jian Liu, Ping Wu, Du-Yun Ye
ABSTRACT
Lipoxin A4 (LXA4), a potent antioxidant and anti-inflammation mediator, protects brains against cerebral ischemia/reperfusion (I/R) injury in vivo. However, few reports concern its function on astrocytes during cerebral I/R injury. The pathogenesis of cerebral I/R injury involves oxidative stress caused by reactive oxygen species (ROS). Upregulation of nuclear factor erythroid 2-related factor 2 (Nrf2) is generally considered to reduce oxidative stress. Nrf2 can induce heme oxygenase-1 (HO-1) expression and glutathione (GSH) release to combat increased oxidative stress. We investigated the effects of LXA4 on astrocytic cell damage, the production of ROS, and Nrf2 pathway, especially on HO-1 expression and GSH release in cultured cortical astrocytes exposed to oxygen-glucose deprivation (OGD)/recovery (OGDR) insults. Primary astrocytes were subjected to a 4-h OGD, followed by 8-h recovery. Cell viability, the production of ROS, and GSH release were measured. Furthermore, Nrf2, HO-1, and p62 expression levels were determined by Western blot. Moreover, Nrf2 location was studied by immunofluorescence staining. Treatment of LXA4 attenuates OGDR-induced cell damage and the production of ROS in a concentration-related manner. LXA4 induced Nrf2 expression and its nuclear translocation, as well as HO-1 expression and GSH release. Moreover, LXA4 induced the excess p62 accumulation. These results indicate that LXA4 can effectively protect against OGDR-induced cell damage in astrocytes, and activation of Nrf2 pathway to reduce oxidative stress may be involved in its protective effects. p62 accumulation induced by LXA4 may be closely related to Nrf2 activation.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein diacetate, used as cell viability stain
Sigma-Aldrich
4,5-Dimethylthiazole, ≥97%, FG
Sigma-Aldrich
DAPI, for nucleic acid staining