Merck
  • Breast Cancer Cell-Derived GM-CSF Licenses Regulatory Th2 Induction by Plasmacytoid Predendritic Cells in Aggressive Disease Subtypes.
Breast Cancer Cell-Derived GM-CSF Licenses Regulatory Th2 Induction by Plasmacytoid Predendritic Cells in Aggressive Disease Subtypes.
Cancer research (2015-05-16)
Cristina Ghirelli, Fabien Reyal, Marine Jeanmougin, Raphaël Zollinger, Philémon Sirven, Paula Michea, Christophe Caux, Nathalie Bendriss-Vermare, Marie-Hélène Donnadieu, Martial Caly, Virginie Fourchotte, Anne Vincent-Salomon, Brigitte Sigal-Zafrani, Xavier Sastre-Garau, Vassili Soumelis
ABSTRACT
Reciprocal interactions between tumor cells and their microenvironment vitally impact tumor progression. In this study, we show that GM-CSF produced by primary breast tumor cells induced the activation of plasmacytoid predendritic cells (pDC), a cell type critical to anti-viral immunity. pDC that expressed the GM-CSF receptor were increased in breast tumors compared with noninvolved adjacent breast tissue. Tumor-activated pDC acquired naïve CD4(+) T-cell stimulatory capacity and promoted a regulatory Th2 response. Finally, the concomitant increase of GM-CSF and pDC was significantly associated with relatively more aggressive breast cancer subtypes. Our results characterize the first tumor-derived factor that can activate pDC to promote a regulatory Th2 response, with implications for therapeutic targeting of a tumor-immune axis of growing recognition in its significance to cancer.
Sigma-Aldrich
Interleukin-3 human, IL-3, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
IL-3 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
IL-3 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Interleukin-3 from mouse, IL-3, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
IL-3 from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
IL-3 from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
IL-3 from mouse, Carrier free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), suitable for cell culture