Merck
  • Nitrate-cancrinite precipitation on quartz sand in simulated Hanford tank solutions.
Nitrate-cancrinite precipitation on quartz sand in simulated Hanford tank solutions.
Environmental science & technology (2002-01-05)
B R Bickmore, K L Nagy, J S Young, J W Drexler
ABSTRACT
Caustic NaNO3 solutions containing dissolved Al were reacted with quartz sand at 89 degrees C to simulate possible reactions between leaked nuclear waste and primary subsurface minerals at the U.S. Department of Energy's Hanford site in Washington. Nitrate-cancrinite began to precipitate onto the quartz after 2-10 days, cementing the grains together. Estimates of the equilibrium constant for the precipitation reaction differ for solutions with 0.1 or 1.0 m OH- (log Keq = 30.4 +/- 0.8 and 36.2 +/- 0.6, respectively). The difference in solubility may be attributable to more perfect crystallinity (i.e., fewer stacking faults) in the higher-pH cancrinite structure. This is supported by electron micrographs of crystal morphology and measured rates of Na volatilization under an electron beam. Precipitate crystallinity may affect radionuclide mobility, because stacking faults in the cancrinite structure can diminish its zeolitic cation exchange properties. The precipitation rate near the onset of nucleation depends on the total Al and Si concentrations in solution. The evolution of experimental Si concentrations was modeled by considering the dependence of quartz dissolution rate on AI(OH)4- activity, cancrinite precipitation, and the reduction of reactive surface area of quartz due to coverage by cancrinite.
Sigma-Aldrich
Sand, white quartz, purum p.a., powder
Pricing and availability is not currently available.
Sigma-Aldrich
Sand, 50-70 mesh particle size
Pricing and availability is not currently available.