Merck
  • Faecalibacterium prausnitzii inhibits interleukin-17 to ameliorate colorectal colitis in rats.
Faecalibacterium prausnitzii inhibits interleukin-17 to ameliorate colorectal colitis in rats.
PloS one (2014-10-03)
Mingming Zhang, Xinyun Qiu, Hao Zhang, Xiaotong Yang, Na Hong, Yonghua Yang, Hui Chen, Chenggong Yu
ABSTRACT
It has been shown that Faecalibacterium prausnitzii (F. prausnitzii), one of the dominant intestinal bacterial flora, may protect colonic mucosa against the development of inflammation and subsequent inflammatory bowel disease (IBD), with the underlying mechanisms being unclear. The impacts of F. prausnitzii and its metabolites on IL-23/Th17/IL-17 pathway markers were determined in human monocytes and a rat model of colitis induced by 2,4,6-trinitrobenzene sulfonic acid. F. prausnitzii and its culture medium (containing complete metabolites) were used to treat the rats in vivo, as well as rat splenocytes and human monocytes in vitro. Inflammatory cytokines were measured in colon tissue, plasma and cell culture medium. The culture supernatant of F. prausnitzii increased plasma anti-Th17 cytokines (IL-10 and IL-12)and suppressed IL-17 levels in both plasma and colonic mucosa, with ameliorated colonic colitis lesions. This inhibition of IL-17 release has also been observed in both rat splenocytes and human venous monocytes in vitro. The culture supernatant of F. prausnitzii also suppressed Th17 cell differentiation induced by cytokines (TGF-ß and IL-6) and bone marrow-derived dendritic cells (BMDCs) in vitro. The metabolites of F. prausnitzii in the culture supernatant exert a stronger anti-inflammatory effect than the bacterium itself. F. prausnitzii protected the colon mucosa against the development of IBD by its metabolites, suggesting a promising potential for the use of F. prausnitzii and its metabolic products in the treatment of IBD.
Sigma-Aldrich
Interleukin-6 from rat, IL-6, recombinant, expressed in E. coli, buffered aqueous solution, suitable for cell culture
Sigma-Aldrich
Interleukin-4 from rat, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥97% (SDS-PAGE)
Sigma-Aldrich
Granulocyte-Macrophage Colony-Stimulating Factor from rat, GM-CSF, recombinant, expressed in E. coli
Sigma-Aldrich
IL-6 from rat, recombinant, expressed in E. coli, ≥90% (SDS-PAGE), ≥90% (HPLC)
Sigma-Aldrich
Picrylsulfonic acid solution, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines