Merck
  • Activated human mast cells induce LOX-1-specific scavenger receptor expression in human monocyte-derived macrophages.
Activated human mast cells induce LOX-1-specific scavenger receptor expression in human monocyte-derived macrophages.
PloS one (2014-09-25)
Mervi Alanne-Kinnunen, Jani Lappalainen, Katariina Öörni, Petri T Kovanen
ABSTRACT
Activated mast cells in atherosclerotic lesions degranulate and release bioactive compounds capable of regulating atherogenesis. Here we examined the ability of activated human primary mast cells to regulate the expression of the major scavenger receptors in cultured human primary monocyte-derived macrophages (HMDMs). Components released by immunologically activated human primary mast cells induced a transient expression of lectin-like oxidized LDL receptor (LOX-1) mRNA in HMDMs, while the expression of two other scavenger receptors, MSR1 and CD36, remained unaffected. The LOX-1-inducing secretory components were identified as histamine, tumor necrosis factor alpha (TNF-α), and transforming growth factor beta (TGF-β1), which exhibited a synergistic effect on LOX-1 mRNA expression. Histamine induced a transient expression of LOX-1 protein. Mast cell -induced increase in LOX-1 expression was not associated with increased uptake of oxidized LDL by the macrophages. Mast cell-derived histamine, TNF-α, and TGF-β1 act in concert to induce a transient increase in LOX-1 expression in human primary monocyte-derived macrophages. The LOX-1-inducing activity potentially endows mast cells a hitherto unrecognized role in the regulation of innate immune reactions in atherogenesis.
Sigma-Aldrich
Goat Anti-Human IgE Antibody, 3.1 mg/mL, Chemicon®
Sigma-Aldrich
Histamine, ≥97.0%
Supelco
Histamine, analytical standard