Merck
  • Evaluation of human D-amino acid oxidase inhibition by anti-psychotic drugs in vitro.
Evaluation of human D-amino acid oxidase inhibition by anti-psychotic drugs in vitro.
Bioscience trends (2014-07-18)
Miho Shishikura, Hitomi Hakariya, Sumiko Iwasa, Takashi Yoshio, Hideaki Ichiba, Kazuko Yorita, Kiyoshi Fukui, Takeshi Fukushima
ABSTRACT
It is of importance to determine whether antipsychotic drugs currently prescribed for schizophrenia exert D-amino acid oxidase (DAO)-inhibitory effects. We first investigated whether human (h)DAO can metabolize D-kynurenine (D-KYN) to produce the fluorescent compound kynurenic acid (KYNA) by using high-performance liquid chromatography with mass spectrometry, and fluorescence spectrometry. After confirmation of KYNA production from D-KYN by hDAO, 8 first- and second-generation antipsychotic drugs, and 6 drugs often prescribed concomitantly, were assayed for hDAO-inhibitory effects by using in vitro fluorometric methods with D-KYN as the substrate. DAO inhibitors 3-methylpyrazole-5-carboxylic acid and 4H-thieno[3,2-b]pyrrole-5-carboxylic acid inhibited KYNA production in a dose-dependent manner. Similarly, the second-generation antipsychotics blonanserin and risperidone were found to possess relatively strong hDAO-inhibitory effects in vitro (5.29 ± 0.47 μM and 4.70 ± 0.17 μM, respectively). With regard to blonanserin and risperidone, DAO-inhibitory effects should be taken into consideration in the context of their in vivo pharmacotherapeutic efficacy.
Sigma-Aldrich
Quetiapine hemifumarate salt, ≥98% (HPLC)
Sigma-Aldrich
Aripiprazole, ≥98% (HPLC)
Quetiapine for system suitability, European Pharmacopoeia (EP) Reference Standard
Aripiprazole, European Pharmacopoeia (EP) Reference Standard
Quetiapine fumarate, European Pharmacopoeia (EP) Reference Standard