Merck
  • Inflammatory bowel disease: MR- and SPECT/CT-based macrophage imaging for monitoring and evaluating disease activity in experimental mouse model--pilot study.
Inflammatory bowel disease: MR- and SPECT/CT-based macrophage imaging for monitoring and evaluating disease activity in experimental mouse model--pilot study.
Radiology (2014-01-31)
Yingwei Wu, Karen Briley-Saebo, Jingyuan Xie, Ruihua Zhang, Zhongqiu Wang, Cijiang He, Cheuk Ying Tang, Xiaofeng Tao
ABSTRACT
To evaluate the feasibility of using magnetic resonance (MR) imaging and single photon emission computed tomography (SPECT)/computed tomography (CT) to visualize the in vivo recruitment of iron oxide-labeled macrophages and indium 111 ((111)In)-labeled macrophages in inflammatory bowel disease (IBD) and to monitor disease activity. This study had institutional animal care and use committee approval. Twenty-seven C57/B6 mice with dextran sodium sulfate (DSS)-induced IBD and control mice were included. Peritoneal macrophages were harvested from seven thioglycollate-treated mice and were labeled with superparamagnetic iron oxide (SPIO) nanoparticles. Macrophage iron content was determined by using inductively coupled plasma mass spectrometry. SPIO nanoparticle-labeled macrophages (5 × 10(6)) were intravenously administered. Mice with DSS-induced IBD (n = 8) and control mice (n = 6) were imaged with a 9.4-T MR imaging unit at 0, 5, and 24 hours after macrophage administration. Percentage normalized enhancement (NE) was calculated for the intestinal wall and liver 24 hours after injection. Six mice with IBD coinjected with SPIO nanoparticles and (111)In oxine-labeled macrophages were imaged with MR imaging and SPECT/CT after 24 hours. The pharmacokinetics and biodistribution of the implanted macrophages were determined. Correlation between percentage NE and IBD scores was calculated. Ex vivo mass spectrometry revealed strong SPIO nanoparticle uptake (7.4 pg iron per cell). R2* correlated with cell number (r = 0.9813, P < .05). Percentage NE correlated with both clinical (r = 0.924) and pathologic (r = 0.795) IBD score. Cell circulation half-life in the first and second phases was 0.32 hour and 10.2 hours, respectively. SPECT/CT showed that approximately 3% of the injected dose was present in the intestines 24 hours after injection; this was confirmed at MR imaging and histologic examination. Indium 111-labeled cells were present in all tissue associated with the reticuloendothelial system or mononuclear phagocyte system at 24 hours. SPIO nanoparticles and (111)In-labeled macrophages could be observed in vivo at MR imaging and SPECT/CT in mice with IBD. Percentage NE at MR imaging correlates with disease activity.
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~100,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 60,000-76,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 15,000-25,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~60,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 48,000-90,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 150,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000
Sigma-Aldrich
Dextran solution from Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)
Dextran 4 for calibration, Dextran 4 European Pharmacopoeia (EP) Reference Standard
Dextran 1, Dextran 1 European Pharmacopoeia (EP) Reference Standard
Dextran Vo, Dextran Vo European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~200,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~40,000
Sigma-Aldrich
Dextran, enzymatic synth.
Supelco
Dextran, analytical standard, 25,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 50,000
Sigma-Aldrich
Magnesium chloride solution, 1.00 M±0.01 M
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 410,000
Supelco
Dextran, analytical standard, Set Mp 1,000-400,000
Supelco
Dextran, analytical standard, 270,000
Supelco
Dextran, analytical standard, 12,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 5,000