Merck
  • Determination of organochlorine pesticide residues in fatty foods: a critical review on the analytical methods and their testing capabilities.
Determination of organochlorine pesticide residues in fatty foods: a critical review on the analytical methods and their testing capabilities.
Journal of chromatography. A (2011-07-12)
Stephen W C Chung, Benedict L S Chen
ABSTRACT
Organochlorine pesticide (OCP) residues in foods have been of concern for several decades. However, the analysis of some of the OCPs and their metabolites or derivatives, such as endrin aldehyde, endrin ketone, nonachlor, etc. in fatty foods (including foods of animal and plant origin), was not commonly included in routine monitoring programme. Recently, the Stockholm Convention introduced nine plus one new persistent organic pollutants (POPs) that included chlordecone and some other OCPs. Is there a method available that can analyze both traditional OCPs, together with their metabolites and derivatives in fatty foods? Furthermore, is there a suitable method that can monitor OCPs and the newly added POPs including chlordecone in fatty foods together in a pot? This review aims to provide some background information to answer these questions.
Product Description
Supelco
Lipid Removal Agent (LRA), pkg of 100 g
Supelco
Lipid Removal Agent (LRA), pkg of 500 g