Merck
Drug-induced permeabilization of S. cerevisiae.
Current protocols in molecular biology (2010-10-05)
Maria D Mayan
ABSTRACT
Budding yeast are not permeable to many drugs. This unit provides a protocol in which polygodial is used to permeabilize the cell membrane, thereby allowing budding yeast cells to be treated with drugs that otherwise would be ineffective.
Sigma-Aldrich
Polygodial, ≥97% (HPLC)