Merck
  • GTDC2 modifies O-mannosylated α-dystroglycan in the endoplasmic reticulum to generate N-acetyl glucosamine epitopes reactive with CTD110.6 antibody.
GTDC2 modifies O-mannosylated α-dystroglycan in the endoplasmic reticulum to generate N-acetyl glucosamine epitopes reactive with CTD110.6 antibody.
Biochemical and biophysical research communications (2013-09-18)
Mitsutaka Ogawa, Naosuke Nakamura, Yoshiaki Nakayama, Akira Kurosaka, Hiroshi Manya, Motoi Kanagawa, Tamao Endo, Koichi Furukawa, Tetsuya Okajima
ABSTRACT
Hypoglycosylation is a common characteristic of dystroglycanopathy, which is a group of congenital muscular dystrophies. More than ten genes have been implicated in α-dystroglycanopathies that are associated with the defect in the O-mannosylation pathway. One such gene is GTDC2, which was recently reported to encode O-mannose β-1,4-N-acetylglucosaminyltransferase. Here we show that GTDC2 generates CTD110.6 antibody-reactive N-acetylglucosamine (GlcNAc) epitopes on the O-mannosylated α-dystroglycan (α-DG). Using the antibody, we show that mutations of GTDC2 identified in Walker-Warburg syndrome and alanine-substitution of conserved residues between GTDC2 and EGF domain O-GlcNAc transferase resulted in decreased glycosylation. Moreover, GTDC2-modified GlcNAc epitopes are localized in the endoplasmic reticulum (ER). These data suggested that GTDC2 is a novel glycosyltransferase catalyzing GlcNAcylation of O-mannosylated α-DG in the ER. CTD110.6 antibody may be useful to detect a specific form of GlcNAcylated O-mannose and to analyze defective O-glycosylation in α-dystroglycanopathies.
Sigma-Aldrich
N-Acetyl-D-glucosamine, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-D-glucosamine, ≥99% (HPLC)
Sigma-Aldrich
N-Acetyl-D-glucosamine, ≥95% (HPLC)
Sigma-Aldrich
Anti-C3ORF39 antibody produced in rabbit, IgG fraction of antiserum