Merck
  • A safety study of thallium-containing ceramic superconductor material in rats.
A safety study of thallium-containing ceramic superconductor material in rats.
Veterinary and human toxicology (1998-02-19)
J P Mulkey, F W Oehme
ABSTRACT
Thallium is a highly toxic cumulative poison in humans and animals. The use of the metal as a component in ceramic superconductor material (CSM) raised concern about the health risk of CSM to children accidentally swallowing some of the CSM or to occupationally-exposed workers in the electronics industry. This study examined the biological availability of CSM by quantifying the thallium content in organs, blood and fecal matter of Sprague-Dawley rats after a single acute oral exposure to CSM and compares these values to similar thallium measurements from identical exposure to thallium sulfate. The CSM-exposed group had significantly less thallium than the thallium sulfate-exposed group in all tissues/fluids analyzed. This suggested that CSM poses a lower health risk to exposed persons than thallium sulfate.
Sigma-Aldrich
Thallium(I) sulfate, 99.99% trace metals basis
Sigma-Aldrich
Thallium(I) sulfate, ≥99.9% trace metals basis