Merck
Electronic cigarette: a possible substitute for cigarette dependence.
Monaldi archives for chest disease = Archivio Monaldi per le malattie del torace (2013-06-08)
P Caponnetto, C Russo, C M Bruno, A Alamo, M D Amaradio, R Polosa
ABSTRACT
Cigarette smoking is the leading cause of premature mortality in western countries and it is important for smokers to stop as early as possible. Electronic cigarettes are a popular phenomenon of global proportion. Recent uncontrolled studies, reported that a certain number of smokers have quit using electronic cigarettes. This could hint a role for electronic cigarettes to be used for smoking cessation, and therefore merits further evaluation for this purpose. Besides vaporising nicotine to be inhaled, electronic cigarettes may also provide a coping mechanism for conditioned smoking cues by replacing some of the rituals associated with smoking gestures, and for these reasons cigarette could become a tool--if studied more extensively--in the fight against tobacco-related morbidity and mortality.
Product Description
Supelco
S(−)-Nicotine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
(−)-Nicotine solution, 1.0 mg/mL, analytical standard, for drug analysis
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Supelco
(−)-Nicotine, PESTANAL®, analytical standard