Merck
  • Modulation of mGluR-dependent MAP1B translation and AMPA receptor endocytosis by microRNA miR-146a-5p.
Modulation of mGluR-dependent MAP1B translation and AMPA receptor endocytosis by microRNA miR-146a-5p.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2013-05-24)
Ying-Lin Chen, Che-Kun James Shen
ABSTRACT
The translation of dendritic microtubule-associated protein 1B (MAP1B) is exaggerated upon group I mGluR activation leading to AMPA receptor (AMPAR) endocytosis and consequent long-term depression. However, the mechanisms of regulation of MAP1B protein synthesis in the mature dendrites remain unclear. Here we have identified miR-146a-5p that targets the 3' UTR of MAP1B mRNA and represses its translation. Inhibition of the endogenous miR-146a-5p in mouse cultured hippocampal neurons triggers an increase of the dendritic MAP1B protein as well as the internalization of AMPARs, resulting in a decline in synaptic transmission. Conversely, enforced expression of miR-146a-5p inhibits MAP1B translation and attenuates group I mGluR-induced AMPAR endocytosis. Moreover, siRNA-mediated knockdown of MAP1B recovers the impairment of synaptic transmission caused by inhibition of miR-146a-5p. These results reveal that miR-146a-5p modulates the synaptic plasticity via repression of MAP1B protein synthesis.
Sigma-Aldrich
DL-3,4-Dihydroxyphenyl glycol
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
DL-2-Amino-5-phosphonopentanoic acid, solid
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
CNQX disodium salt hydrate, ≥98% (HPLC), solid
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
SAFC
L-Valine
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material