Merck
  • Ultrafast dynamics of aniline following 269-238 nm excitation and the role of the S2(pi3s/pi sigma) state.
Ultrafast dynamics of aniline following 269-238 nm excitation and the role of the S2(pi3s/pi sigma) state.
Faraday discussions (2012-12-13)
Roman Spesyvtsev, Oliver M Kirkby, Helen H Fielding
ABSTRACT
Femtosecond time-resolved photoelectron imaging is employed to investigate ultrafast electronic relaxation in aniline, a prototypical aromatic amine. The molecule is excited at wavelengths between 269 and 238 nm. We observe that the S2(pi3s/pi sigma*) state is populated directly during the excitation process at all wavelengths and that the population bifurcates to two decay pathways. One of these involves ultrafast relaxation from the Rydberg component of S2(pi3s/pi sigma*) to the S1(Pi Pi)* state, from which it relaxes back to the electronic ground state on a much longer timescale. The other appears to involve motion along the pi sigma* dissociative potential energy surface. At higher excitation energies, the dominant excitation is to the S3(pi pi*) state, which undergoes extremely efficient electronic relaxation back to the ground state. Our study supports some conclusions reached from H-atom photofragment translational spectroscopy measurements and pump-probe photoionization measurements and contradicts some others.
Sigma-Aldrich
Aniline-4-13C, 99 atom % 13C
Sigma-Aldrich
Aniline-15N, 98 atom % 15N
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Supelco
Aniline, analytical standard
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sigma-Aldrich
Aniline, SAJ first grade, ≥99.0%
Sigma-Aldrich
Aniline, JIS special grade, ≥99.0%
Sigma-Aldrich
Aniline-1-13C, 99 atom % 13C
Supelco
Aniline solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Aniline hydrochloride, ≥99%