Merck
  • Live birth after SrCl(2) oocyte activation in previous repeated failed or low fertilization rates after ICSI of frozen-thawed testicular spermatozoa: case report.
Live birth after SrCl(2) oocyte activation in previous repeated failed or low fertilization rates after ICSI of frozen-thawed testicular spermatozoa: case report.
Journal of assisted reproduction and genetics (2012-11-28)
Jun-Woo Kim, Jung-Lim Choi, Seong-Ho Yang, San-Hyun Yoon, Jae-Hoon Jung, Jin-Ho Lim
ABSTRACT
To report a live birth resulting after strontium chloride (SrCl(2)) oocyte activation in a couple with complete fertilization failure or low fertilization rates following intracytoplasmic sperm injection (ICSI) of frozen-thawed testicular spermatozoa. The couple underwent ICSI of frozen-thawed testicular spermatozoa. After ICSI, the oocytes were artificially activated by SrCl(2) because the results of fertilization were not satisfactory in the previous cycles. The main outcome measures were fertilization, pregnancy, and birth. In the first and second cycles performed previously at another clinic, fertilization rates were 9.1 % and 0.0 %, respectively. In the third cycle, 31 metaphase II oocytes were retrieved. After sperm injection, all of the oocytes were stimulated using SrCl(2) for activation. Sixteen oocytes were fertilized (51.6 %), and a single embryo was transferred into the uterus on Day 3. A healthy girl weighing 2750 g was born at 40 weeks of gestation by caesarean section. This result suggests that SrCl(2) could be useful for oocyte fertilization in case of repeated complete fertilization failure or low fertilization rates following ICSI of frozen-thawed testicular spermatozoa.
Sigma-Aldrich
Strontium chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Strontium chloride solution, 0.1 M
Sigma-Aldrich
Strontium chloride hexahydrate, SAJ first grade, ≥98.0%
Sigma-Aldrich
Strontium chloride hexahydrate, 99.995% trace metals basis
Sigma-Aldrich
Strontium chloride hexahydrate, ACS reagent, 99%