Merck
  • The ionic atmosphere around A-RNA: Poisson-Boltzmann and molecular dynamics simulations.
The ionic atmosphere around A-RNA: Poisson-Boltzmann and molecular dynamics simulations.
Biophysical journal (2012-03-06)
Serdal Kirmizialtin, Alexander R J Silalahi, Ron Elber, Marcia O Fenley
ABSTRACT
The distributions of different cations around A-RNA are computed by Poisson-Boltzmann (PB) equation and replica exchange molecular dynamics (MD). Both the nonlinear PB and size-modified PB theories are considered. The number of ions bound to A-RNA, which can be measured experimentally, is well reproduced in all methods. On the other hand, the radial ion distribution profiles show differences between MD and PB. We showed that PB results are sensitive to ion size and functional form of the solvent dielectric region but not the solvent dielectric boundary definition. Size-modified PB agrees with replica exchange molecular dynamics much better than nonlinear PB when the ion sizes are chosen from atomistic simulations. The distribution of ions 14 Å away from the RNA central axis are reasonably well reproduced by size-modified PB for all ion types with a uniform solvent dielectric model and a sharp dielectric boundary between solvent and RNA. However, this model does not agree with MD for shorter distances from the A-RNA. A distance-dependent solvent dielectric function proposed by another research group improves the agreement for sodium and strontium ions, even for shorter distances from the A-RNA. However, Mg(2+) distributions are still at significant variances for shorter distances.
Sigma-Aldrich
Strontium chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Strontium chloride solution, 0.1 M
Sigma-Aldrich
Strontium chloride hexahydrate, SAJ first grade, ≥98.0%
Sigma-Aldrich
Strontium chloride hexahydrate, 99.995% trace metals basis
Sigma-Aldrich
Strontium chloride hexahydrate, ACS reagent, 99%