Merck
  • Effect of solvent annealing on the tensile deformation mechanism of a colloidal crystalline polymeric latex film.
Effect of solvent annealing on the tensile deformation mechanism of a colloidal crystalline polymeric latex film.
Langmuir : the ACS journal of surfaces and colloids (2011-08-31)
Jianqi Zhang, Zhiyong Yi, Qiao Wang, Zhenyu Liu, Jan Perlich, Rainer Gehrke, Yongfeng Men
ABSTRACT
The influence of solvent annealing on microscopic deformational behavior of a styrene/n-butyl acrylate copolymer latex film subjected to uniaxial tensile deformation was studied by small-angle X-ray scattering. It was demonstrated that the microscopic deformation mechanism of the latex films transformed from a nonaffine deformation behavior to an affine deformation behavior after solvent annealing. This was attributed to the interdiffusion of polymeric chains between adjacent swollen latex particles in the film. It turns out that solvent annealing is much more efficient than thermal annealing due to a much slow evaporation process after solvent annealing.
Sigma-Aldrich
Butyl acrylate, ≥99%, contains 10-60 ppm monomethyl ether hydroquinone as inhibitor
Supelco
Butyl acrylate, analytical standard
Sigma-Aldrich
Butyl acrylate, SAJ first grade, ≥99.0%