Merck
  • Acute lethal toxicity, hyperkalemia associated with renal injury and hepatic damage after intravenous administration of cadmium nitrate in rats.
Acute lethal toxicity, hyperkalemia associated with renal injury and hepatic damage after intravenous administration of cadmium nitrate in rats.
Journal of occupational health (2007-02-23)
Emi Dote, Tomotaro Dote, Hiroyasu Shimizu, Yukari Shimbo, Michiko Fujihara, Koichi Kono
ABSTRACT
Cadmium nitrate Cd(NO(3))(2) (CdN) is commonly used in Ni-Cd battery factories. The possibility of accidental exposure to CdN is great. CdN is very soluble in water compared to other Cd compounds. Therefore, acute toxicity would be expected to be quick due to rapid absorption after exposure. However, the mechanisms of CdN toxicity have not been fully elucidated. We investigated the acute lethal toxicity and harmful systemic effects of acute exposure to large doses of CdN. The lethal dose and dose-response study of the liver and kidney were determined after intravenous administration of CdN in rats. The LD(50) of CdN was determined to be 5.5 mg/kg. Doses of 2.1, 4.2, 6.3 mg/kg were selected for the dose-response study. Liver injury was induced at doses greater than 4.2 mg/kg. Severe hepatic injury occurred in the 6.3 mg/kg group, which would have been caused by acute exposure to the high concentration of Cd that exceeded the critical concentration in hepatic tissue. A remarkable decrease in urine volume in the 6.3 mg/kg group indicated acute renal failure. A decrease in creatinine clearance suggested acute glomerular dysfunction at doses greater than 4.2 mg/kg. Increases in urinary N-acetyl-beta-D-glucosaminidase/creatinine, beta(2)-microglobulin and glucose in the 6.3 mg/kg group indicated proximal tubular injury. Secretion of K ion was also severely affected by proximal tubular injury and severe decreases in urine volume, and an increase in serum K ion was identified at doses greater than 4.2 mg/kg. Thus severe hyperkalemia might be associated with the cardiac-derived lethal toxicity of CdN.
Sigma-Aldrich
Cadmium nitrate tetrahydrate, 99.997% trace metals basis
Sigma-Aldrich
Cadmium nitrate tetrahydrate, 98%
Sigma-Aldrich
Cadmium standard solution, suitable for atomic absorption spectrometry, 1 mg/mL Cd+2, 1000 ppm Cd+2